Assay Details
Gene Symbol: JPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003639124.3 | XP_003639172.1 | 1 - 2 | 119 | 115 | |
XM_022420273.1 | XP_022275981.1 | 1 - 2 | 119 | 115 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003639124.3, XM_022420273.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
