Assay Details
| Gene Symbol: JPT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639124.3 | XP_003639172.1 | 2 - 3 | 270 | 72 | |
| XM_022420273.1 | XP_022275981.1 | 2 - 3 | 270 | 72 | ||
More Information
Set Membership: |
|

JPT2
Genomic Map