Assay Details
| Gene Symbol: CCDC59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014119530.2 | XP_013975005.1 | 1 - 2 | 297 | 78 | |
| XM_022428135.1 | XP_022283843.1 | 1 - 2 | 297 | 78 | ||
| XM_849268.5 | XP_854361.1 | 1 - 2 | 297 | 78 | ||
More Information
Set Membership: |
|

CCDC59
Genomic Map