Assay Details
| Gene Symbol: WDR41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014112255.2 | XP_013967730.1 | 1124 | 59 | ||
| XM_014112257.2 | XP_013967732.1 | 1040 | 59 | |||
| XM_014112258.2 | XP_013967733.1 | 980 | 59 | |||
| XM_022416765.1 | XP_022272473.1 | 1181 | 59 | |||
| XM_022416766.1 | XP_022272474.1 | 1166 | 59 | |||
| XM_022416767.1 | XP_022272475.1 | 1118 | 59 | |||
| XM_022416768.1 | XP_022272476.1 | 1118 | 59 | |||
More Information
Set Membership: |
|

WDR41
Genomic Map