Assay Details
| Gene Symbol: RILPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005636118.3 | XP_005636175.1 | 845 | 59 | ||
| XM_848133.5 | XP_853226.2 | 833 | 59 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: RILPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005636118.3 | XP_005636175.1 | 845 | 59 | ||
| XM_848133.5 | XP_853226.2 | 833 | 59 | |||
Set Membership: |
|