Assay Details
| Gene Symbol: TMEM247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005626237.3 | XP_005626294.1 | 1 - 2 | 214 | 76 | |
| GenBank mRNA | CX987277.1 | - | 2 - 3 | 517 | 76 | |
| DN744266.1 | - | 1 - 2 | 151 | 76 | ||
More Information
Set Membership: |
|

TMEM247
Genomic Map