Assay Details
| Gene Symbol: RGS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005622405.3 | XP_005622462.1 | 568 | 68 | ||
| XM_005622407.3 | XP_005622464.1 | 442 | 68 | |||
| XM_014115122.2 | XP_013970597.1 | 381 | 68 | |||
| XM_022420953.1 | XP_022276661.1 | 510 | 68 | |||
| XM_022420954.1 | XP_022276662.1 | 397 | 68 | |||
| XM_022420955.1 | XP_022276663.1 | 375 | 68 | |||
| XM_844457.5 | XP_849550.1 | 296 | 68 | |||
More Information
Set Membership: |
|

RGS8
Genomic Map