Assay Details
| Gene Symbol: UPK3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005620991.3 | XP_005621048.1 | 842 | 76 | ||
| XM_022419591.1 | XP_022275299.1 | 684 | 76 | |||
| XM_844184.5 | XP_849277.3 | 819 | 76 | |||
More Information
Set Membership: |
|

UPK3B
Genomic Map