Assay Details
| Gene Symbol: NUPR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003434817.4 | XP_003434865.1 | 1 - 2 | 329 | 63 | |
| XM_536921.6 | XP_536921.2 | 1 - 2 | 321 | 63 | ||
More Information
Set Membership: |
|

NUPR1
Genomic Map