Assay Details
| Gene Symbol: DLA-DMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048099.1 | NP_001041564.1 | 3 - 4 | 517 | 69 | |
| XM_005627068.3 | XP_005627125.1 | 5 - 6 | 1403 | 69 | ||
| XM_014118041.2 | XP_013973516.1 | 4 - 5 | 822 | 69 | ||
| XM_014118042.2 | XP_013973517.1 | 4 - 5 | 799 | 69 | ||
More Information
Set Membership: |
|

DLA-DMA
Genomic Map