Assay Details
Gene Symbol: KRT13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001346369.1 | NP_001333298.1 | 750 | 80 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KRT13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001346369.1 | NP_001333298.1 | 750 | 80 |