Assay Details
| Gene Symbol: FAM32A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287071.1 | NP_001274000.1 | 4 - 5 | 368 | 75 | |
| GenBank mRNA | DN417984.1 | - | 1 - 2 | 363 | 75 | |
More Information
Set Membership: |
|

FAM32A
Genomic Map