Assay Details
| Gene Symbol: PCGF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014108937.2 | XP_013964412.1 | 1 - 2 | 439 | 191 | |
| XM_022411895.1 | XP_022267603.1 | 1 - 2 | 439 | 191 | ||
| XM_846928.5 | XP_852021.1 | 1 - 2 | 434 | 191 | ||
More Information
Set Membership: |
|

PCGF6
Genomic Map