Assay Details
| Gene Symbol: ABHD17B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005615858.2 | XP_005615915.1 | 3 - 4 | 671 | 129 | |
| XM_022421173.1 | XP_022276881.1 | 3 - 4 | 1065 | 129 | ||
| XM_541286.6 | XP_541286.2 | 3 - 4 | 1302 | 129 | ||
More Information
Set Membership: |
|

ABHD17B
Genomic Map