Assay Details
Gene Symbol: FHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003435535.4 | XP_003435583.2 | 4 - 5 | 664 | 134 | |
XM_005641862.3 | XP_005641919.1 | 4 - 5 | 669 | 134 | ||
XM_014111636.2 | XP_013967111.1 | 5 - 6 | 619 | 134 | ||
XM_022415593.1 | XP_022271301.1 | 5 - 6 | 619 | 134 | ||
XM_861215.5 | XP_866308.3 | 4 - 5 | 648 | 134 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
