Assay Details
| Gene Symbol: LDB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003433613.5 | XP_003433661.1 | 8 - 9 | 924 | 94 | |
| XM_005637646.3 | XP_005637703.1 | 8 - 9 | 739 | 94 | ||
| XM_022411685.1 | XP_022267393.1 | 8 - 9 | 763 | 94 | ||
| XM_022411687.1 | XP_022267395.1 | 8 - 9 | 752 | 94 | ||
| XM_845833.5 | XP_850926.2 | 8 - 9 | 739 | 94 | ||
More Information
Set Membership: |
|

LDB1
Genomic Map