Assay Details
| Gene Symbol: STEAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005628486.3 | XP_005628543.1 | 4 - 5 | 1540 | 140 | |
| XM_539408.6 | XP_539408.3 | 4 - 5 | 1651 | 140 | ||
More Information
Set Membership: |
|

STEAP2
Genomic Map