Assay Details
| Gene Symbol: PRELID2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005617340.3 | XP_005617397.1 | 8 - 9 | 910 | 187 | |
| XM_014108174.2 | XP_013963649.1 | 7 - 8 | 768 | 187 | ||
| XM_014108195.2 | XP_013963670.1 | 7 - 8 | 699 | 187 | ||
| XM_014108196.2 | XP_013963671.1 | 8 - 9 | 1070 | 187 | ||
| XM_014108202.2 | XP_013963677.1 | 7 - 8 | 765 | 187 | ||
| XM_014108213.2 | XP_013963688.1 | 6 - 7 | 626 | 187 | ||
| XM_022409062.1 | XP_022264770.1 | 8 - 9 | 1001 | 187 | ||
| XM_022409071.1 | XP_022264779.1 | 5 - 6 | 425 | 187 | ||
| XM_022409079.1 | XP_022264787.1 | 8 - 9 | 841 | 187 | ||
| XM_022409082.1 | XP_022264790.1 | 8 - 9 | 907 | 187 | ||
| XM_853408.5 | XP_858501.1 | 5 - 6 | 613 | 187 | ||
More Information
Set Membership: |
|

PRELID2
Genomic Map