Assay Details
| Gene Symbol: ZGRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014110061.2 | XP_013965536.1 | 28 - 29 | 6427 | 173 | |
| XM_022413411.1 | XP_022269119.1 | 28 - 29 | 6516 | 173 | ||
| XM_022413412.1 | XP_022269120.1 | 27 - 28 | 6367 | 173 | ||
| XM_022413413.1 | XP_022269121.1 | 27 - 28 | 6346 | 173 | ||
| XM_022413414.1 | XP_022269122.1 | 27 - 28 | 6240 | 173 | ||
| XM_022413415.1 | XP_022269123.1 | 26 - 27 | 6181 | 173 | ||
| XM_022413416.1 | XP_022269124.1 | 26 - 27 | 6177 | 173 | ||
| XM_022413417.1 | XP_022269125.1 | 29 - 30 | 5858 | 173 | ||
| XM_022413418.1 | XP_022269126.1 | 28 - 29 | 5828 | 173 | ||
More Information
Set Membership: |
|

ZGRF1
Genomic Map