Assay Details
| Gene Symbol: ANKRD54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014117367.2 | XP_013972842.1 | 4 | 621 | 91 | |
| XM_022424461.1 | XP_022280169.1 | 3 | 522 | 91 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ANKRD54
Genomic Map