Assay Details
| Gene Symbol: STOML2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022425546.1 | XP_022281254.1 | 4 - 5 | 458 | 110 | |
| XM_531986.6 | XP_531986.2 | 4 - 5 | 458 | 110 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

STOML2
Genomic Map