Assay Details
| Gene Symbol: PDZRN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639968.4 | XP_003640016.1 | 3 - 4 | 708 | 81 | |
| XM_022411099.1 | XP_022266807.1 | 3 - 4 | 483 | 81 | ||
| XM_022411100.1 | XP_022266808.1 | 3 - 4 | 462 | 81 | ||
| XM_022411101.1 | XP_022266809.1 | 3 - 4 | 384 | 81 | ||
More Information
Set Membership: |
|

PDZRN4
Genomic Map