Assay Details
| Gene Symbol: ART5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014122571.2 | XP_013978046.1 | 3 | 1823 | 119 | |
| XM_014122572.2 | XP_013978047.1 | 3 | 1401 | 119 | ||
| XM_022407740.1 | XP_022263448.1 | 3 | 1728 | 119 | ||
| XM_022407741.1 | XP_022263449.1 | 3 | 1823 | 119 | ||
More Information
Set Membership: |
|

ART5
Genomic Map