Assay Details
| Gene Symbol: LAYN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005619797.3 | XP_005619854.2 | 563 | 162 | ||
| XM_014113427.2 | XP_013968902.2 | 566 | 162 | |||
| XM_022418327.1 | XP_022274035.1 | 575 | 162 | |||
More Information
Set Membership: |
|

LAYN
Genomic Map