Assay Details
| Gene Symbol: BMP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197052.1 | NP_001183981.1 | 1 | 253 | 69 | |
| XM_005634690.3 | XP_005634747.1 | 1 | 371 | 69 | ||
More Information
Set Membership: |
|

BMP7
Genomic Map