Assay Details
| Gene Symbol: JADE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005626544.3 | XP_005626601.2 | 12 | 2546 | 119 | |
| XM_005626545.3 | XP_005626602.1 | 12 | 2769 | 119 | ||
| XM_005626546.3 | XP_005626603.1 | 13 | 2955 | 119 | ||
| XM_005626547.3 | XP_005626604.1 | 13 | 2732 | 119 | ||
| XM_005626548.2 | XP_005626605.1 | 13 | 2731 | 119 | ||
| XM_005626549.3 | XP_005626606.1 | 12 | 2678 | 119 | ||
| XM_005626550.3 | XP_005626607.1 | 10 | 2356 | 119 | ||
| XM_005626551.3 | XP_005626608.1 | 12 | 2542 | 119 | ||
| XM_022425313.1 | XP_022281021.1 | 10 | 2448 | 119 | ||
| XM_845623.5 | XP_850716.3 | 12 | 2547 | 119 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

JADE2
Genomic Map