Assay Details
| Gene Symbol: TNMD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022416193.1 | XP_022271901.1 | 5 - 6 | 855 | 111 | |
| XM_538101.5 | XP_538101.1 | 6 - 7 | 951 | 111 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TNMD
Genomic Map