Assay Details
| Gene Symbol: TTBK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005638315.3 | XP_005638372.1 | 6 - 7 | 819 | 86 | |
| XM_005638316.3 | XP_005638373.1 | 5 - 6 | 569 | 86 | ||
| XM_005638317.3 | XP_005638374.1 | 5 - 6 | 524 | 86 | ||
| XM_022412910.1 | XP_022268618.1 | 6 - 7 | 645 | 86 | ||
| XM_022412911.1 | XP_022268619.1 | 6 - 7 | 605 | 86 | ||
| XM_544644.6 | XP_544644.3 | 6 - 7 | 741 | 86 | ||
More Information
Set Membership: |
|

TTBK2
Genomic Map