Assay Details
| Gene Symbol: SGCD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005619257.3 | XP_005619314.1 | 2 - 3 | 480 | 133 | |
| XM_014113050.2 | XP_013968525.1 | 3 - 4 | 462 | 133 | ||
| XM_014113051.2 | XP_013968526.1 | 2 - 3 | 211 | 133 | ||
| XM_022417768.1 | XP_022273476.1 | 2 - 3 | 481 | 133 | ||
More Information
Set Membership: |
|

SGCD
Genomic Map