Assay Details
| Gene Symbol: MPO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022423192.1 | XP_022278900.1 | 7 - 8 | 896 | 125 | |
| XM_022423193.1 | XP_022278901.1 | 7 - 8 | 885 | 125 | ||
| XM_847352.4 | XP_852445.2 | 7 - 8 | 893 | 125 | ||
More Information
Set Membership: |
|

MPO
Genomic Map