Assay Details
| Gene Symbol: SERPINA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005641622.2 | XP_005641679.1 | 4 | 942 | 178 | |
| XM_014111361.2 | XP_013966836.1 | 4 | 1058 | 178 | ||
| XM_538128.5 | XP_538128.3 | 3 | 900 | 178 | ||
More Information
Set Membership: |
|

SERPINA7
Genomic Map