Assay Details
| Gene Symbol: GABRD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005620414.2 | XP_005620471.1 | 382 | 71 | ||
| XM_005620416.3 | XP_005620473.1 | 167 | 71 | |||
| XM_014113862.2 | XP_013969337.1 | 297 | 71 | |||
| XM_546729.5 | XP_546729.3 | 380 | 71 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005620416.3 transcripts.
Set Membership: |
|

GABRD
Genomic Map