Assay Details
| Gene Symbol: UBE2W | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022412441.1 | XP_022268149.1 | 3 - 4 | 307 | 140 | |
| XM_022412442.1 | XP_022268150.1 | 3 - 4 | 307 | 140 | ||
| XM_544128.5 | XP_544128.3 | 3 - 4 | 307 | 140 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

UBE2W
Genomic Map