Assay Details
| Gene Symbol: ANKRD34B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005618168.3 | XP_005618225.1 | 336 | 128 | ||
| XM_005618169.3 | XP_005618226.1 | 238 | 128 | |||
| XM_022416757.1 | XP_022272465.1 | 1733 | 128 | |||
More Information
Set Membership: |
|

ANKRD34B
Genomic Map