Assay Details
| Gene Symbol: NGF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194950.1 | NP_001181879.1 | 1 | 532 | 161 | |
| GenBank mRNA | AY800386.1 | - | 1 | 413 | 161 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

NGF
Genomic Map