Assay Details
| Gene Symbol: COX15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022412119.1 | XP_022267827.1 | 4 - 5 | 624 | 80 | |
| XM_022412120.1 | XP_022267828.1 | 3 - 4 | 449 | 80 | ||
| XM_022412121.1 | XP_022267829.1 | 3 - 4 | 453 | 80 | ||
| XM_534986.6 | XP_534986.2 | 4 - 5 | 691 | 80 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

COX15
Genomic Map