Assay Details
| Gene Symbol: ETV5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005639799.3 | XP_005639856.1 | 2 - 3 | 287 | 159 | |
| XM_022414125.1 | XP_022269833.1 | 2 - 3 | 422 | 159 | ||
More Information
Set Membership: |
|

ETV5
Genomic Map