Assay Details
| Gene Symbol: EPYC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005629219.3 | XP_005629276.1 | 6 - 7 | 902 | 113 | |
| XM_014119758.2 | XP_013975233.1 | 7 - 8 | 1021 | 113 | ||
| XM_014119759.2 | XP_013975234.1 | 7 - 8 | 996 | 113 | ||
More Information
Set Membership: |
|

EPYC
Genomic Map