Assay Details
| Gene Symbol: KCNJ8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014108709.2 | XP_013964184.1 | 2 | 800 | 141 | |
| XM_543766.6 | XP_543766.2 | 3 | 618 | 141 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

KCNJ8
Genomic Map