Assay Details
| Gene Symbol: MORF4L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003435631.4 | XP_003435679.1 | 4 | 838 | 118 | |
| XM_003435632.4 | XP_003435680.1 | 4 | 787 | 118 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MORF4L2
Genomic Map