Assay Details
| Gene Symbol: LHX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003435342.3 | XP_003435390.1 | 3 - 4 | 442 | 59 | |
| XM_005625407.2 | XP_005625464.1 | 3 - 4 | 441 | 59 | ||
| XM_005625408.3 | XP_005625465.1 | 3 - 4 | 321 | 59 | ||
More Information
Set Membership: |
|

LHX6
Genomic Map