Assay Details
| Gene Symbol: RIMS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005628872.3 | XP_005628929.1 | 4 | 682 | 121 | |
| XM_844171.5 | XP_849264.1 | 4 | 685 | 121 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: RIMS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005628872.3 | XP_005628929.1 | 4 | 682 | 121 | |
| XM_844171.5 | XP_849264.1 | 4 | 685 | 121 | ||
Set Membership: |
|