Assay Details
| Gene Symbol: ZNF652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005624627.3 | XP_005624684.1 | 1475 | 77 | ||
| XM_022423095.1 | XP_022278803.1 | 1546 | 77 | |||
| XM_022423096.1 | XP_022278804.1 | 1170 | 77 | |||
More Information
Set Membership: |
|

ZNF652
Genomic Map