Assay Details
| Gene Symbol: CCZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022419722.1 | XP_022275430.1 | 1018 | 153 | ||
| XM_536878.6 | XP_536878.5 | 1014 | 153 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CCZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022419722.1 | XP_022275430.1 | 1018 | 153 | ||
| XM_536878.6 | XP_536878.5 | 1014 | 153 | |||
Set Membership: |
|