Assay Details
| Gene Symbol: RASD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003434639.4 | XP_003434687.1 | 1 | 134 | 86 | |
| XM_546661.6 | XP_546661.2 | 1 | 134 | 86 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: RASD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003434639.4 | XP_003434687.1 | 1 | 134 | 86 | |
| XM_546661.6 | XP_546661.2 | 1 | 134 | 86 | ||
Set Membership: |
|