Assay Details
| Gene Symbol: CLDN18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022408667.1 | XP_022264375.1 | 1 | 319 | 144 | |
| XM_534274.5 | XP_534274.2 | 1 | 306 | 144 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CLDN18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022408667.1 | XP_022264375.1 | 1 | 319 | 144 | |
| XM_534274.5 | XP_534274.2 | 1 | 306 | 144 | ||
Set Membership: |
|