Assay Details
| Gene Symbol: ZNF771 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621287.2 | XP_005621344.1 | 452 | 115 | ||
| XM_014114378.2 | XP_013969853.1 | 525 | 115 | |||
More Information
Set Membership: |
|

ZNF771
Genomic Map