Assay Details
| Gene Symbol: RNF43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014116698.2 | XP_013972173.1 | 6 - 7 | 1498 | 107 | |
| XM_014116699.2 | XP_013972174.1 | 5 - 6 | 567 | 107 | ||
| XM_014116702.2 | XP_013972177.1 | 7 - 8 | 804 | 107 | ||
| XM_022423197.1 | XP_022278905.1 | 6 - 7 | 692 | 107 | ||
| XM_022423198.1 | XP_022278906.1 | 4 - 5 | 454 | 107 | ||
More Information
Set Membership: |
|

RNF43
Genomic Map