Assay Details
| Gene Symbol: RNPEP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022420804.1 | XP_022276512.1 | 625 | 113 | ||
| XM_022420805.1 | XP_022276513.1 | 364 | 113 | |||
| XM_547357.6 | XP_547357.2 | 623 | 113 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_022420805.1 transcripts.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RNPEP
Genomic Map