Assay Details
| Gene Symbol: ZBTB26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005625397.2 | XP_005625454.1 | 2 | 1371 | 113 | |
| XM_005625398.3 | XP_005625455.1 | 2 | 1361 | 113 | ||
| XM_014117081.2 | XP_013972556.1 | 2 | 1386 | 113 | ||
| XM_022423874.1 | XP_022279582.1 | 2 | 1291 | 113 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ZBTB26
Genomic Map